Would you like to log in again?
Coating Thickness Tester, Extech
Supplier: Extech
CG206
75989-452EA 622.25 USD InStock
75989-452
Coating Thickness Tester, Extech
Coating Testers
Dual technology provides automatic recognition for ferrous and non-ferrous substrates without having you guess the material.
  • Smart automatic substrate recognition
  • Magnetic induction for ferrous substrates
  • Eddy current measurement for non-ferrous substrates
  • Two working modes: direct and group
  • Memory stores 1500 readings (30 group readings)
  • Two point calibration with zero adjust
  • USB interface includes software
Allows for quick, one-handed operation, non- destructive measurement of the coating thickness on various surfaces.
Certifications: CE Certified.
Ordering information: Includes 2 AAA batteries, USB cable, software, Calibration Iron, Calibration Aluminum, Precision Standards, and pouch.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty