Would you like to log in again?
Nylon Ace-Thred Bushing Connector, Ace Glass Incorporated
Supplier: Ace Glass
7506-50
75877-644EA 48.74 USD InStock
75877-644
Nylon Ace-Thred Bushing Connector, Ace Glass Incorporated
Fittings and Connectors
Nylon bushing connector for joining an Ace-Thred to a reduced-end tube.
Nylon bushing connector is supplied with a FETFE O-ring and and Ace-Thred tap. FETFE is a fluoroelastomer with special TFE additives that has a good compression set along with excellent temperature and chemical compatibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty