Would you like to log in again?
Ward's® Essentials Properties of Polymers Lab Kit
Supplier: ALDON CORP SE
Ward's®
AD-WE2505-KIT
470225-524EA 65.79 USD InStock
470225-524
Ward's® Essentials Properties of Polymers Lab Kit
Educational Classroom Kits and Activities
Learn about and create three types of polymers and explore the differences.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-PS1-2, MS-PS1-3
  • Instructions and materials included for 15 groups
  • Middle school
Students will create polymer worms, polymer slime, and polymer snow as they explore differences between thermoplastic and thermoset polymers. Students will also investigate properties of various crosslinked, superabsorbent polymers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty