Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palmitic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: Hexadecanoic acidHexadecylic acid

101600100MG
100293-562EA 99.38 USD InStock
100293-562
Palmitic acid ≥99%
Palmitic acid
Store in a cool, dry place


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
MDL Number: MFCD00002747
CAS Number: 57-10-3
ADR: ,
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty