Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Nonane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

050900100MG
100293-044EA 115.08 USD InStock
100293-044
n-Nonane ≥99%
n-Nonane
Store in a cool, dry place


Formula: H₃C(CH₂)₇CH₃
MW: 128.26 g/mol
MDL Number: MFCD00009574
CAS Number: 111-84-2
UN: 1920
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty