Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triolein ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: Glycerol TrioleateGlycerine trioleate

331810100MG 331810500MG
100278-300EA 255 USD InStock
100278-300 104018-212
Triolein ≥99%
Triolein
Store in a cool, dry place


Formula: C₅₇H₁₀₄O₆
MW: 885.45 g/mol
MDL Number: MFCD00137563
CAS Number: 122-32-7
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty