Would you like to log in again?
Tergitol™ NP-4, Surfactant, Spectrum Chemical
Supplier: Spectrum
Tergitol™
T1276-100ML T1276-500MLG T1276-4LTGL T1276-20LT
700011-136EA 100 USD InStock
700011-136 700011-132 700011-134 700011-130
Tergitol™ NP-4, Surfactant, Spectrum Chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ NP-4 is a Nonylphenol Ethoxylate surfactant. It is non-ionic and provides excellent detergency, rinseability, and low odor. This product is suggested for use in cleaners & degreasers, pre-wash spotters and metal-working fluids.
  • Excellent oil-soluble surfactant
  • Low HLB emulsifier
  • Low odor
  • Exceptional handling properties
  • Insoluble in water
Density at 20°C (68°F): 1.027 g/ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty