Would you like to log in again?
Sample Pans for Therman Analyzers, PerkinElmer
Supplier: Perkin Elmer
N5200045
10136-612PK 3489.35 USD InStock
10136-612
Sample Pans for Therman Analyzers, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
These ceramic sample pan kits are used in thermal analyzers, including the STA 6000 and TGA 4000.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty