Would you like to log in again?
High Performance Polyester Labels, Type B30, WorkHorse®, Brady
Supplier: Brady
B30
143584
10018-040EA 260.68 USD InStock
10018-040
High Performance Polyester Labels, Type B30, WorkHorse®, Brady
Labels
High Performance Polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces, including stainless steel and polypropylene.
  • Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Ideal for general and barcode labeling on flat, smooth surfaces
Labels are good chemical resistance, best for application to smoother surfaces indoors. Enables printers with XY cutters to cut custom shapes or multiple labels out of a single 4" continuous roll wide material using BBP®37 Printer.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty