Would you like to log in again?
High Temperature Polyimide Labels, Brady
Supplier: Brady
B30
145297
10018-128EA 1963.78 USD InStock
10018-128
High Temperature Polyimide Labels, Brady
Labels
High Temperature Polyimide (B-727) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labeling.
  • Ultra durable adhesive is designed for use with extreme wash protocol and cleaning chemicals, and is ideal for use in auto apply equipment
  • Glossy white polyimide material is thermal transfer printable
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Certifications: RoHS compliant to 2011/65/EU; UL approved; Meets requirements of MIL-STD-202G, Method 215K
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty