Would you like to log in again?
Permanent Polyester Labels, Type B30, WorkHorse®, Brady
Supplier: Brady
B30
143578
10017-986EA 285.15 USD InStock
10017-986
Permanent Polyester Labels, Type B30, WorkHorse®, Brady
Labels
Permanent polyester label (B-423) is ideal for electronic PCB and component identification, barcode labels, rating plates, and solar panel identification.
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.
Meets requirements of a halogen-free material per DIN VDE 0472 part 815.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty