Would you like to log in again?
SP Wilmad-LabGlass Stainless Steel Inlet Filters, SP Industries
Supplier: SP Industries
BP-1916-000 BP-1916-621
14007-328EA 63.94 USD InStock
14007-328 14007-334
SP Wilmad-LabGlass Stainless Steel Inlet Filters, SP Industries
Filters Liquid Process Filters
Inlet solvent filters are economical and provide optimal protection by filtering solvents to prevent pump cavitation and other system component damage.
  • Disposable filters are easy to replace; simply tighten by hand
  • Wetted parts are stainless steel
  • Filters are ultra-high molecular weight polyethylene (UHMWPE)
The 2 µm porosity filter is designed for use with ¹/₈" ext.Ø. tubing. The 10 µm porosity filter is supplied with an ETFE finger-tight fitting cap (male nut) with a ¹/₄"-28 thread port configuration, and is designed for use with ¹/₈" ext.Ø tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty