Would you like to log in again?
Ready-to-Assay™ D5 Dopamine Receptor Frozen Cells, Eurofins Pharma Discovery Services
Supplier: Eurofins Pharma Discovery Services
HTS129RTA
103382-982PK 1013.85 USD InStock
103382-982
Ready-to-Assay™ D5 Dopamine Receptor Frozen Cells, Eurofins Pharma Discovery Services
Assays Cellular Assays
Human D5 GPCR frozen cells for Calcium Flux FLIPR Assays.
  • Calcium Flux Assay
  • cAMP Accumulation
  • Host Chem-1
  • Adherent rat hematopoietic cell line
  • Expresses endogenous Gα15 protein
Dopamine is a catecholamine neurotransmitter that functions in the CNS to control locomotor, cognitive, emotional and neurendocrine processes, and in the periphery to modulate cardiovascular, renal and gastrointestinal processes. The biological activities of dopamine are mediated by a family of five GPCRs. The D1 and D5 subtypes couple to Gs to increase intracellular cAMP, whereas the D2, D3 and D4 subtypes couple to Gi to reduce cAMP (Missale et al., 1998). The hypertensive phenotype of mice with a targeted deletion of D5 indicates that D5 regulates central control of sympathetic vascular tone (Hollon et al., 2002). In addition, D5 modulates locomotion and corticostriatal long-term depression (Centonze et al., 2003). Cloned human D5-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant D5 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at D5.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty