Would you like to log in again?
Brady® PRINZING® Economy Polyester Lockout Tagout Tags, Brady Worldwide
Supplier: Brady
45655
82004-268EA 39.38 USD InStock
82004-268
Brady® PRINZING® Economy Polyester Lockout Tagout Tags, Brady Worldwide
Security Locks
Economy Polyester (B-851) tags are perfect for marking on-the-spot warnings of dangerous conditions.
  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb. pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates, and other special instructions
  • Withstands moisture, grease, and dirt
  • Stays legible longer than cardstock
Certifications: ANSI Z53.1-1967. ANSI Z535-2006. OSHA 1910.145. OSHA 1910.147.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty