Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nα-Boc-Nε-trifluoroacetyl-L-lysine ≥99% (by HPLC), white powder

Supplier: CHEM IMPEX INTL INC MS
Synonyms: N2-Boc-N6-(2,2,2-trifluoroacetyl)-L-lysine

03802-5G 03802-1G 03802-25G
102638-854EA 386.43 USD InStock
102638-854 102618-602 102586-484
Nα-Boc-Nε-trifluoroacetyl-L-lysine ≥99% (by HPLC), white powder
N-α-Boc-N-φ-trifluoroacetyl-L-lysine


Formula: C₁₃H₂₁F₃N₂O₅
MW: 342.315 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037104
CAS Number: 16965-06-3
Order Now
Specification Test Results
Appearance White powder
Solubility Clear solution (0.3 g in 2 mL DMF)
Water Content (Karl Fisher) ≤1.0%
Optical Rotation -11 ± 1º (C=2 in DMF)
Infrared Spectrum Conforms to structure
Analysis by Mass Spectra Conforms to standard
Loss on Drying ≤1.0% (50 °C, 2 h)
1H NMR Conforms to structure
Purity by HPLC ≥99%
Storage Store at 0 - 8 °C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty