Would you like to log in again?
LB Lennox Agar, Teknova
Supplier: Teknova
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
L9330 L9335
200066-620EA 788.33 USD InStock
200066-620 101320-276
LB Lennox Agar, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of E. coli and other enteric bacteria.
  • Add 35 grams per 1 Liter of water to reconstitute
  • Autoclave to sterilize
  • Ambient shipping conditions
  • Store at room temperature
LB Lennox contains less salt than most common formulations of LB, known as LB Miller
Ingredients: 1.0% Tryptone (10.0g/L), 0.5% Yeast Extract (5.0g/L), 0.5% NaCl (5.0g/L), and 1.5% Agar (15.0g/L).
LB Lennox is commonly used for cloning in recombinant E. coli and selection with salt-sensitive antibiotics such as Blasticidin, CloNat, Hygromycin, Puromycin and Zeocin.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty