Would you like to log in again?
SiliaSep™ Imidazole Flash Cartridges, SiliCycle
Supplier: SiliCycle
FR79230BIS750 FR79230BI1500 FR79230BISO40 FR79230BISO80 FR79230BISO04 FR79230BIS220 FR79230BIS120 FR79230BISO25 FR79230BIS750 FR79230BISO25 FR79230BISO12 FR79230BISO12 FR79230BIS220 FR79230BI1500 FR79230BIS330 FR79230BIS120 FR79230BIS330 FR79230BISO40 FR79230BISO04 FR79230BISO80
75880-278BX 1638.22 USD InStock
75880-278 CA75880-276 CA75880-288 CA75880-278 75880-292 CA75880-290 75880-282 CA75880-280 CA75880-292 CA75880-282 75880-284 75880-274 75880-286 CA75880-284 CA75880-274 75880-276 75880-288 CA75880-286 75880-290 75880-280
SiliaSep™ Imidazole Flash Cartridges, SiliCycle
Chromatography Columns
With its SiliaSep™ line, SiliCycle offers pre-packed Flash cartridges of superior performance.
  • Highest silica gel quality
  • Innovative packing technology
  • Universal Luer fittings
  • Compatible with any flash system
  • Cost effective
  • Multiple sorbent variety and cartridge sizes
With a more tightly packed silica bed and a homogeneous packing, the use of pre-packed flash cartridges improves purification efficiency by offering superior reproducibility and productivity compared to conventional manual flash chromatography.
SiliaSep Flash cartridges offers outstanding performance and reliability for all your separation requirements, from milligrams to kilograms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty