Would you like to log in again?
Casein Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G122
75993-532PK 126.05 USD InStock
75993-532
Casein Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of aerobic actinomycetes based on casein proteolysis.
  • Produces clear halo
  • Can hydrolyse casein
Organisms that can hydrolyse casein, such as Streptomyces, Pseudomonas, and Actinomadura spp. Will produce a clear halo in the surrounding medium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty