Would you like to log in again?
HTM Agar (Haemophilus Test Medium) for susceptibility testing, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
H07 G33
75993-536PK 78.65 USD InStock
75993-536 76177-352
HTM Agar (Haemophilus Test Medium) for susceptibility testing, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For disk diffusion sensitivity testing of Haemophilus spp
by the Kirby-Bauer method.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty