Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
XLT4 Agar / Brilliant Green Agar with Novobiocin, Hardy Diagnostics
Supplier: Hardy Diagnostics
J127BX
75993-472PK 327.43 USD InStock
75993-472
XLT4 Agar / Brilliant Green Agar with Novobiocin, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and differentiation of Salmonella spp
from environmental samples and food.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty