Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HardyDisks™ Ampicillin/Sulbactam, SAM-20, Hardy Diagnostics
Supplier: Hardy Diagnostics
HardyDisks™
Z8045
75993-456EA 87.26 USD InStock
75993-456
HardyDisks™ Ampicillin/Sulbactam, SAM-20, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty