Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Flavivirus Recombinant Monoclonal Antibody [clone: D1-4G2-4-15 (4G2)]

Supplier: BioVision
AB286196-200UG
76005-718EA 896.32 USD InStock
76005-718
Anti-Flavivirus Recombinant Monoclonal Antibody [clone: D1-4G2-4-15 (4G2)]
Antibodies
This antibody binds to flavivirus group antigen, protein E. It can be used as an anti-Dengue virus antibody, anti-West Nile virus antibody, anti-Japanese Encephalitis or anti-Zika Virus antibody (Aubry et al. 2016) to identify cells infected with these flaviviridae. It binds to the fusion loop at the extremity of domain II of E protein from all four serotypes and prevents syncytia formation. The epitope is highly conserved amongst flaviviridae and has been functionally analyzed in detail by Crill and Chang 2004. Previous studies have used acetone- (Henchal et al. 1982) or methanol-fixed slides (Moreland & Tay, 2010). Please note that binding of this antibody has been reported to be sensitive to reduction and Western Blots should be performed under non-reducing conditions (Lai et al. 2008).

Type: Primary
Antigen: Flavivirus
Clonality: Monoclonal
Clone: D1-4G2-4-15 (4G2)
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty