Would you like to log in again?
Human Recombinant Glyceraldehyde-3-phosphate dehydrogenase (active) (from E. coli)
Supplier: BioVision
AB291253-10UG AB291253-50UG
76006-240EA 160.88 USD InStock
76006-240 76006-242
Human Recombinant Glyceraldehyde-3-phosphate dehydrogenase (active) (from E. coli)
Proteins and Peptides
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a catalytic enzyme commonly known to be involved in glycolysis. The enzyme exists as a tetramer composed of 36-kDa subunits and has various intracellular functions. GAPDH catalyzes the reversible reduction of 1,3-bisphosphoglycerate to glyceraldehyde 3-phosphophate in the presence of NADPH. Besides functioning as a glycolytic enzyme in cytoplasm, evidence suggests that mammalian GAPDH is also involved in a great number of intracellular processes such as membrane fusion, microtubule bundling, phosphotransferase activity, nuclear RNA export, DNA replication and DNA repair. Recombinant GAPDH protein was expressed in E.coli and purified by using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty