Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD4 Monoclonal Antibody (R-PE) [clone: M-T441]

Supplier: AdipoGen Life Sciences
ANC-148-050
102511-916EA 443.08 USD InStock
102511-916
Anti-CD4 Monoclonal Antibody (R-PE) [clone: M-T441]
Antibodies
Human CD4 is a cell surface glycoprotein of 55kDa expressed on most thymocytes, on about two thirds of peripheral T cells and on some monocyte macrophage lineage cells. CD4 interacts with MHC Class II molecules in an accessory role during foreign antigen recognition by T cells. The extracellular portion of CD4 is an array of four Ig-like domains. The N-terminal region of CD4 is a receptor for the HIV-1 viral protein gp120.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: M-T441
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG2b
Reactivity: Baboon, Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty