Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD14 Monoclonal Antibody (FITC) [clone: UCHM1]

Supplier: AdipoGen Life Sciences
ANC-163-040
102512-050EA 346.15 USD InStock
102512-050
Anti-CD14 Monoclonal Antibody (FITC) [clone: UCHM1]
Antibodies
Human CD14 is strongly expressed on myelomonocyte lineage cells including monocytes, macrophages and a subset of granulocytes. CD14 is a high affinity receptor for complexes of lipopolysaccharide and lipopolysaccharide binding protein. A soluble form of CD14 also binds to LPS and LBP.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: UCHM1
Conjugation: FITC
Epitope:
Host:
Isotype: Mouse IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty