Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Croton oil

Supplier: Enzo Life Sciences
Danger
Synonyms: Tiglium oil

350-089-KI225
76001-992EA 3690 USD InStock
76001-992
Croton oil
Croton oil
Natural source of phorbol and phorbol esters.

Purified from Croton tiglium L.

Soluble in DMSO, 100% ethanol or acetone. Insoluble in cold water.


Storage Temperature: Refrigerator
MDL Number: MFCD00130880
CAS Number: 8001-28-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty