Would you like to log in again?
AlphaRsaAlphaI
Supplier: Enzo Life Sciences
GEN109-1000
76003-290EA 78.69 USD InStock
76003-290
AlphaRsaAlphaI
Proteins and Peptides
Produced in an E. coli strain that carries the cloned RsaI gene from Rhodopseudomonas sphaeroides.
  • High quality enzyme with stringent QC testing
  • Includes 10X Cutting Buffer
  • BSA premixed into buffers
RsaI is a restriction enzyme that recognizes GT^AC sites.10X Cutting Buffer is included that contains BSA, which enhances enzyme stability and binds to contaminants in DNA preps.
Suitable for molecular cloning, restriction site mapping, genotyping, southern blotting, SNP.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty