Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FLUOR DE LYS®-succinyl green, desuccinylase substrate 5 mM in DMSO (synthetic) , 95% (by HPLC)

Supplier: Enzo Life Sciences
Warning

BML-KI591-0050
76002-386EA 245.72 USD InStock
76002-386
FLUOR DE LYS®-succinyl green, desuccinylase substrate 5 mM in DMSO (synthetic) , 95% (by HPLC)
FLUOR DE LYS®-Succinyl Green, Desuccinylase Substrate
The FLUOR DE LYS®-Succinyl green, desuccinylase substrate offers a highly sensitive and convenient alternative to radiolabeled, succinylated histones or peptide/HPLC methods for the assay of desuccinylases such as SIRT5.

The substrate has longer wavelength excitation and emission. This avoids interference by quenching or fluorescence from compounds absorbing and emitting in the near UV and blue range.

Can be used for the study of desuccinylase kinetics, regulation, and inhibitor sensitivity. Ideal for drug discovery and HTS applications.


Order Now
Specification Test Results
Concentration 5 mM
Source Synthetic
Absorbance maximum 470 - 500 nm
Emission maximum 520 - 550 nm
Purity ≥95% (HPLC)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty