Would you like to log in again?
MMP-3 fluorogenic Substrate
Supplier: Enzo Life Sciences
BML-P278-0100
76002-136EA 246.66 USD InStock
76002-136
MMP-3 fluorogenic Substrate
Proteins and Peptides
Long emission wavelength reduces background
5'-FAM-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(TQ2W)-NH2
Highly-quenched, ultra-bright fluorogenic substrate for MMP-3 (other MMPs not tested, but likely).  5’-FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMP-3 separates the two moieties.
Highly-quenched, ultra-bright fluorogenic substrate for MMP-3 (other MMPs not tested, but likely).  5’-FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMP-3 separates the two moieties.
5'-FAM-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(TQ2W)-NH2
Highly-quenched, ultra-bright fluorogenic substrate for MMP-3 (other MMPs not tested, but likely).  5’-FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMP-3 separates the two moieties.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty