Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SUMO1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0505A-0100 PW0505A-0025
76002-144EA 152.05 USD InStock
76002-144 76002-390
Anti-SUMO1 Rabbit Polyclonal Antibody
Antibodies
Recommended Applications: ICC, IP, WB

Species reactivity: Human

Type: Primary
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty