Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HTT Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0595A-0100 PW0595A-0025
76002-398EA 505.74 USD InStock
76002-398 76002-396
Anti-HTT Rabbit Polyclonal Antibody
Antibodies
Recommended Applications: WB

Species reactivity: Mouse, Rat

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty