Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]

Supplier: Enzo Life Sciences
ABS120-1000 ABS120-0200 ABS120-0500
76002-614EA 549.55 USD InStock
76002-614 76002-616 76002-618
Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]
Antibodies
Recommended Applications: IF, IHC, IP, WB

Species reactivity:

Type: Primary
Antigen: HA-tag
Clonality: Monoclonal
Clone: 16B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Hamster, Cat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty