Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Mouse Monoclonal Antibody [clone: B34]

Supplier: Enzo Life Sciences
ABS141-0200
76002-672EA 521.69 USD InStock
76002-672
Anti-GFP Mouse Monoclonal Antibody [clone: B34]
Antibodies
Recommended Applications: WB

Species reactivity:

Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: B34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty