Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD39 Mouse Monoclonal Antibody [clone: AC2.5]

Supplier: Enzo Life Sciences
ABS233-0100
76002-820EA 602.47 USD InStock
76002-820
Anti-CD39 Mouse Monoclonal Antibody [clone: AC2.5]
Antibodies
Recommended Applications: Flow Cytometry, IHC (FS), IP

Species reactivity: Human

Type: Primary
Antigen: CD39
Clonality: Monoclonal
Clone: AC2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty