Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAS9 Mouse Monoclonal Antibody [clone: 7A9]

Supplier: Enzo Life Sciences
ABS294-0100
76002-938EA 572.74 USD InStock
76002-938
Anti-CAS9 Mouse Monoclonal Antibody [clone: 7A9]
Antibodies
Recommended Applications: IF, IP, WB

Species reactivity:

Type: Primary
Antigen: CAS9
Clonality: Monoclonal
Clone: 7A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty