Would you like to log in again?
10K Spin Column, Biovision Inc.
Supplier: BioVision
AB93349-25
10013-838EA 632.97 USD InStock
10013-838
10K Spin Column, Biovision Inc.
Filters Centrifugal Filters
The 10 Kd spin columns are disposable ultrafiltration devices for the concentration of biological samples.
  • Columns to deproteinate or concentrate samples
  • Maximum initial sample volume 500 µl
  • Dead stop volume: ~20 to 25 µl
They can be used in either a swing bucket or fixed angle rotors accepting 2.0 ml centrifuge tube at around 10000 g. The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentration, even laden solutions. Specifically designed with low internal surface and membrane area in order to achieve superior recovery from dilute solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty