Would you like to log in again?
Bovine Native Albumin (from Blood)
Supplier: MP Biomedicals
08820451 08820452
IC820451EA 1163.38 USD InStock
IC820451 76045-160
Bovine Native Albumin (from Blood)
Proteins and Peptides
Plasma undergoes salt fractionation using caprylic acid (sodium salt), heating at 65 °C for three hours; alcohol precipitation followed by an acetone wash resulting in a Fraction V powder.
  • Protease-free
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty