Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C3 Goat Polyclonal Antibody

Supplier: MP Biomedicals
0855444
75983-050EA 500.07 USD InStock
75983-050
Anti-C3 Goat Polyclonal Antibody
Antibodies
The complement system provides innate defense against microbial infection and is a "complement" to antibody mediated immunity. The complement system consists of thirty-five interacting plasma and membrane associated proteins which contribute to host-defense and initiate and amplify inflammation, even in the preimmune state where specific antibodies and lymphocytes are not available. In addition to the complement components themselves, this system also contains several soluble factors that prevent spontaneous complement activation from occurring in solution, as well as, several regulatory membrane associated proteins that protect host cells from accidental complement attack. Cleavage of the C3 component by either the classical pathway or the alternative pathway releases C3a and C3b. C3a appears to be important in many inflammatory responses while the C3b fragment covalently binds to the cell or bacterial surface and plays a role in opsonisation. Binding of C3b to the C4b component of the C3 convertase, results in C5 convertase (C4b3b2a) formation.

Cell staining, tissue staining, immunostaining

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty