Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Red Blood Cells Rabbit Polyclonal Antibody

Supplier: MP Biomedicals
0855800
75983-068EA 179.25 USD InStock
75983-068
Anti-Red Blood Cells Rabbit Polyclonal Antibody
Antibodies
immunoblotting, Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections

Type: Primary
Antigen: Red Blood Cells
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty