Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgE Goat Polyclonal Antibody (Biotin)

Supplier: Tonbo Biosciences
30-8026-M001
76040-310EA 273 USD InStock
76040-310
Anti-IgE Goat Polyclonal Antibody (Biotin)
Antibodies
Goat anti-Human IgE (epsilon chain) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: ε chain
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty