Would you like to log in again?
Antigen Retrieval Reagent, pH 6 (10X), Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC112-0100
75873-994EA 216.41 USD InStock
75873-994
Antigen Retrieval Reagent, pH 6 (10X), Enzo Life Sciences
Protein Extraction Protein Extraction Kits
Antigen Retrieval Reagent, pH 6 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty