Would you like to log in again?
Human Recombinant FGF-4 (from E. coli)
Supplier: BioVision
AB285468-5MG
10834-178EA 37691.21 USD InStock
10834-178
Human Recombinant FGF-4 (from E. coli)
Proteins and Peptides
Fibroblast Growth Factor 4 (FGF-4) is a growth factor predominantly expressed during embryonic development, playing a key role in limb development. In culture, FGF-4 has been shown to be an important growth factor for fibroblasts and endothelial cells. Human FGF-4 shares high homology and cross-reactivity with the mouse protein. Recombinant human FGF-4, produced in E.coli, is a non-glycosylated protein containing 177 amino acids and having a total molecular mass of 19 kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty