Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nourseothricin sulfate ≥85%

Supplier: BioVision
Synonyms: Streptothricin sulfateclonNATStreptothricin sulphateNTC

AB285409-100MG AB285409-50MG AB285409-1G AB285409-500MG
76021-874EA 509.35 USD InStock
76021-874 76021-876 76021-878 76021-872
Nourseothricin sulfate ≥85%
Nourseothricin sulfate
Nourseothricin is group of a broad-spectrum antibiotic produced by Streptomyces variants; inhibits protein synthesis by inducing miscoding

Streptothricin antibiotic Nourseothricin (NTC) is used for a broad spectrum of viruses, bacteria and other unicellular or complex organisms. Preferred selection antibiotic for genetically modified Gram-positive and Gram-negative bacteria, yeast, filamentous fungi, protozoa, plants and many more. Selection of recombinant strains is based on inactivation of NTC by monoacetylation of ß-amino group of the ß-lysine by Nouresothricin N-acetyltransferase, the product of the sat1 gene.

Caution: For Research Use Only! Not For Use in Humans.


Formula: C₃₁H₅₈N₁₂O₁₀·C₁₉H₃₄N₈O₈·H₂SO₄
MW: 1359.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00165013
CAS Number: 96736-11-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/53/23492353.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty