Would you like to log in again?
HeLa Nuclear Extract, Biovision Inc.
Supplier: BioVision
AB286851-1MG AB286851-100UG
10005-234EA 222.15 USD InStock
10005-234 10005-236
HeLa Nuclear Extract, Biovision Inc.
Cells
For western blotting and HDAC assay positice controls.
  • Application- Western blot- 10-20 µg/lane; HDAC Fluorometric Assay- 10 µg/assay; HDAC Colorimetric Assay- 50 µg/assay
Nuclear extract is prepared from HeLa cells using a modified protocol of Dignam et al., which contains a variety of DNA-binding proteins, transcription factors, and other nuclear proteins.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty