Would you like to log in again?
IL-6 (human), High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT178-0001
75840-746EA 807.05 USD InStock
75840-746
IL-6 (human), High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
Most sensitive (0.057 pg/mL) colorimetric kit for IL-6 quantification
  • High-sensitivity ELISA enables quantification of low amounts of IL-6
  • Easy-to-use liquid color-coded reagents reduce errors
  • Rapid quantitative results in 3 hours
  • High-throughput immunoassay measures up to 40 samples in duplicate per 96 well kit
  • Ready-to-use reagents and pre-coated plates reduce errors and save time
For the quantitative determination of human IL-6 in culture supernatants, plasma, serum, and urine.
The IL-6 (human), high sensitivity ELISA kit is a colorimetric immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. Measure up to 40 samples in duplicate per 96 well  kit.
Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty