Would you like to log in again?
Human Recombinant PPARgamma (from E. coli)
Supplier: BioVision
AB285987-100UG AB285987-500UG AB285987-20UG
75838-034EA 275.79 USD InStock
75838-034 75838-036 75838-038
Human Recombinant PPARgamma (from E. coli)
Proteins and Peptides
The peroxisome proliferator activated receptor (PPAR) family of ligand-activated transcription factors consists of three subtypes encoded by separate genes: PPARα, PPARδ and PPARγ. PPARγ plays an important role in the regulation of fatty acid storage and glucose metabolism.
The genes activated by PPARγ stimulate lipid uptake and adipogenesis by fat cells. Many naturally occurring molecules such as, polyunsaturated fatty acids like arachidonic acid and arachidonic acid metabolites, are known to bind and activate PPARγ. The binding of activating ligands to the ligand binding domain (LBD) of PPARγ promotes its heterodimerization with retinoic acid-like receptor (RXR), which results in the regulated expression of target genes involved in lipid metabolism. BioVision’s His-PPARγ (LBD) is a non-glycosylated protein expressed and purified from E. coli. It has been tested for functionality by its binding and subsequent displacement of 1,8-ANS assay.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty