Would you like to log in again?
AEC Substrate Buffer, Biovision Inc.
Supplier: BioVision
AB286857-50ML
10005-252EA 252.78 USD InStock
10005-252
AEC Substrate Buffer, Biovision Inc.
Buffers Microscopy Buffers
A diluent for 3-Amino-9-ethylcarbazole.
  • APPLICATIONS- Immunoblotting, Immunohistochemistry
A diluent for use with 3-Amino-9-ethylcarbazole (AEC) (Cat.# 2122-10), a precipitable peroxidase substrate for immunoblotting and immunohistochemical staining techniques, supplied as a 10X concentrate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty