Would you like to log in again?
Human Recombinant MMP-14 (from E. coli)
Supplier: BioVision
AB285993-50UG AB285993-10UG
75838-042EA 1133.82 USD InStock
75838-042 75838-040
Human Recombinant MMP-14 (from E. coli)
Enzymes
The Matrix metalloproteinase-14 (MMP-14) belongs to a family of zinc-dependent endopeptidases. The ability of MMP-14 to degrade type I collagen, and activate pro-MMP-2 and pro-MMP-9 makes it a key enzyme in many physiological and pathological processes such as angiogenesis and tumor invasion.
  • Gene ID: 4323
  • Accession number: P50281
MMP-14 is structurally a multi-domain metalloenzyme, composed of a prodomain, a metal-binding catalytic domain, a hinge region, a hemopexin-like domain, a transmembrane domain, and a cytoplamasic tail. The catalytic, and hinge domains of MT1-MMP were expressed in Escherichia coli and the active form was generated by successive autoproteolysis of the N- and C-terminal sites during refolding.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty