Would you like to log in again?
EZ-Desalt™ Spin Desalting Columns, Biovision Inc.
Supplier: BioVision
AB288149-25UNITS
10009-000EA 288.87 USD InStock
10009-000
EZ-Desalt™ Spin Desalting Columns, Biovision Inc.
Chromatography Columns
Columns with pre-swollen Sephadex G-25 beads for buffer exchange and desalting of protein samples.
  • Quickly desalt, buffer-exchange, or clean-up protein samples in solution with exceptional (>90%) recovery rate.
EZ-Desalt™ Spin Desalting Column offers a convenient option to quickly desalt, buffer-exchange, or clean-up protein samples in solution. Each of the disposable, ready-to-use, spin desalting column contains 0.5 ml pre-swollen Sephadex G-25 beads, that efficiently separates proteins (or other high molecular weight components) from small molecules (salt and other small molecules such as LMW dyes etc.). Large molecules are too big to enter the pores of the beads and are excluded from Sephadex matrix and are eluted immediately after the void volume of the column, whereas small molecules are retained by the resin, with the cut-off size of about 7 kDa. The whole process takes few min. and achieves an excellent recovery of proteins (>90%), and efficient removal of the salt or unwanted small molecules.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty