Would you like to log in again?
Mouse Recombinant Sonic Hedgehog (SHH) (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285990-25UG AB285990-100UG AB285990-1MG
75838-086EA 6754.91 USD InStock
75838-086 75838-082 75838-084
Mouse Recombinant Sonic Hedgehog (SHH) (from E. coli)
Proteins and Peptides
Recombinant mouse sonic hedgehog is a member of a small group of secreted proteins that are essential for development in both vertebrates and invertebrates. Three mammalian hedgehog genes (sonic, desert, Indian) share about 60% homology.
The mouse sonic hedgehog is 99% homologous to the human gene. Cysteine at position 25 has been substituted with Ile. Mouse SHH is a non-glycosylated polypeptide consisting of 176 amino acids with a molecular weight of 19800 Da.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty